Protector® XVS Ventilation Stations, Labconco®
This ventilated work area is ideal as a weighing station, light-duty fume hood, science classroom workstation, solvent cleaning work bay, or for veterinary pathology and cytology
The deep 58.4cm (23") interior easily accommodates a variety of equipment including large balances. Glacier white and gray, dry powder epoxy-coated enclosures have a sturdy aluminum frame with 0.5cm (3/16") thick tempered safety glass top and side walls. The angled and hinged sash with 0.6cm (1/4") thick tempered safety glass swings up for easy loading and cleaning. The sash opening on 57.8cm (223/4") high models is 20.3cm (8"); on 81.3cm (32") high models the opening is 25.4cm (10"). The sash pivots upward and locks to a loading height of 50.8cm (20") on 57.8cm (223/4") high models, and 74.3cm (291/4") on the 81.3cm (32") high models.
The Clean-Sweep™ airfoil allows air to sweep the work surface for maximum containment. Airflow openings pull air under the airfoil, creating a constant barrier of protection from contaminants. The upper containment sash foil bleeds air into the enclosure to direct contaminants away from the operator’s breathing zone. The perforated rear baffle promotes horizontal laminar airflow to maximize containment. Additional side-entry airfoils allow air to sweep across the interior glass surfaces to enhance containment. Two utility ports allow passage of tubing and cords. Enclosures feature 5.1x25.4cm (2x10") rectangular exhaust collars on the top and bottom for connection to ductwork or portable exhauster.
Black, chemical-resistant, solid epoxy work surfaces are contoured to fit the dimensions of the enclosure and dished to contain spills. Base stands are made of durable 4.4cm (13/4") thick, tubular, glacier white epoxy-coated steel. The height of base stands is fully adjustable from 70–85cm (271/2–331/2") in 2.5cm (1") increments. Base stands come with either fixed leveling feet for use on uneven floor surfaces or four 12.7cm (5") non-marking casters.
Utility shelf kit includes three epoxy-coated steel shelves that can hold a printer, utensils, and other miscellaneous supplies. Shelves attach to the rear baffle perforations of the enclosure. Remote blowers 30153-980, 30153-982, and 82010-730 have a 1/4hp, totally enclosed, fan-cooled (TEFC) standard motor capable of overcoming external static pressure loss of 0.75in.H2O at 360cfm. Housing and wheel are made of corrosion-resistant, epoxy-coated steel. Blower inlet has an I.D. of 15.2cm (6"). Outlet dimensions are 10.8Wx18.7Lcm (41/4x73/8"). Applications involving high concentrations of solvents may require the explosion-proof blower.
Even, inward airflow protects the operator and contains hazardous or noxious fumes, vapors, and fine powders.
Certifications: All systems conform to ASHRAE 110-1995 standards.
Ordering information: Work surface and stand are not included and must be purchased separately; contact your Avantor representative for more information. Items designated as REDISHIP can be shipped within 48 hours of receiving your order. Enclosures must be connected to either a remote blower or a FilterMate portable exhauster (see 82010-632 series) for proper exhaustion. For Guardian airflow monitors, see 82007-280 series. Gray work surfaces are also available; contact your Avantor sales representative for more information.
Services: VWR CATALYST™ offers customer on-site validation, Testing & certification services including Face Velocity Profile Test, Smoke Generation Test, Tracer Gas Test, IQ, OQ, and PQ. Please call 888-793-2300 or email VWR[email protected] for more details. You can also visit https://vwr.com/equip_services.
Documents
Labconco REDISHIP Literature 2021
Product Literature
- Inside height:311/4"
- Inside width:46¾"
- Outside height:32"
- Outside depth:29"
- Outside width:48"
- Shipping weight:100 lbs.
- Inside depth:23"
- Width:4 ft.
- Exhaust volume:250 cfm at 80 fpm, 185 cfm at 60 fpm
- Rediship:No
Accessories
- DescriptionRedishipShipping weight
- DescriptionRedishipShipping weight
- 30153-980Remote Blower, 115/230V, 50Hz, 5.6/2.8A, 35.6 x 28 x 38.1 cm (14 x 11 x 15")No16 kg (35 lbs.)Each - $1,960.00
- DescriptionRedishipShipping weight
- DescriptionElectricalFor use with
- DescriptionRedishipShipping weight
- DescriptionElectricalFor use with
- DescriptionOutside depthOutside height
- DescriptionShipping weight
- DescriptionInside depthOutside height
- DescriptionFor use withOutside depth
- DescriptionRedishipShipping weight
- DescriptionOutside depthOutside height
- DescriptionOutside depthOutside height
- DescriptionRedishipShipping weight
- Description
- DescriptionRedishipShipping weight
- DescriptionFor use withOutside depth
- DescriptionFor use withInterior dimensions
- DescriptionRedishipShipping weight
- DescriptionRedishipShipping weight
- DescriptionFor use withOutside depth
- DescriptionFor use withInterior dimensions
- DescriptionØHeight
- DescriptionRedishipShipping weight
- DescriptionOutside depthOutside height
- DescriptionElectricalFor use with
- DescriptionRedishipShipping weight
- DescriptionElectricalFor use with
- DescriptionRedishipShipping weight
- Description
- DescriptionFor use withOutside depth
- DescriptionRedishipShipping weight
- DescriptionRedishipShipping weight
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



