Order Entry
United States
ContactUsLinkComponent
PLATE DEEP WELL PP 1.2 ML PK24
BRAND 96-Well and 384-Well Deep Well Plates, BrandTech
Catalog # 80087-066
Supplier:  Brandtech
undefined
BRAND 96-Well and 384-Well Deep Well Plates, BrandTech
Catalog # 80087-066
Supplier:  Brandtech
Supplier Number:  701350

Specifications

  • Sterility:
    Non sterile
  • Material:
    PP
  • Cat. No.:
    80087-066
  • Working volume:
    1.0 ml
  • Supplier no.:
    701350

Specifications

About this item

Deep well plates are excellent for high throughput screening (HTS), assays requiring a mother plate, cell and tissue culture, and sample storage

  • Suitable for storage, reagent reservoirs or use as mother plates
  • Work with most robotic/lquid handling systems
  • Polypropylene modes suitable for –80 °C (–112 °F) cryostorage

Polypropylene plates are manufactured from high-quality virgin polypropylene, are autoclavable to 121 °C (250 °F), and are resistant to a wide variety of chemicals, including phenols, chloroform, and DMSO. Plates are compatible with most robotic samplers and automated liquid handling systems. Polypropylene plates withstand temperatures as low as –80 °C (–112 °F) and are suitable for cryo-storage. Polystyrene plate withstands temperatures as low as –20 °C (–4 °F), making it suitable for cold room applications. Wells are alphanumerically labeled and have notched corners to facilitate orientation in automated systems.

Ordering information: Aluminum sealing film 80087-180 is RNase- and DNase-free and ideal for cryo-storage.
Frequently Bought Together