Orders LinkContactUsLinkComponent
Seals, Optimize Technologies
OPTI-MAX EF Inlet Cartridge Check Valve 3/16" Ceramic Ball & Seat, SS Cartridge, Waters 225 µL (#3) [510, 590, 600/600E, 610, 650, 6K/6KA]
  77101-366
 :   Optimize
 :  39-66-01262
Seals, Optimize Technologies
  77101-366
 :  Optimize
Seals, Optimize Technologies
Seals, Optimize Technologies
OPTI-MAX EF Inlet Cartridge Check Valve 3/16" Ceramic Ball & Seat, SS Cartridge, Waters 225 µL (#3) [510, 590, 600/600E, 610, 650, 6K/6KA]
$778.81
Each
Piston seals are designed to offer reliable performance in a variety of operating conditions.
  • Polyethylene-based seals are designed for applications where mostly aqueous or buffered-aqueous mobile phases are used
  • Graphetized PTFE seals are designed for applications where mobile phases are mostly organic or where chemical compatibility is of primary concern
  • Reliable performance in a variety of operating conditions
Installing a piston seal can be a tricky undertaking. A relatively small seal must be fit into the seal cavity in such a way to ensure that it is pressed in evenly without distortion or damage. The Optimize Seal Forming/ Insertion Tool Kit was specifically designed to aid in this endeavor. This kit contains three tools to accommodate all of your HPLC/ UHPLC piston seals. One end has a beveled cavity for seal forming, while the other is an insertion support for perfect seal installation. Also included with each kit are complete instructions for maximizing seal life and a solvent compatibility guide to aid in prudent seal material selection.
OPTI-SEAL® UHMW-PE polyethylene-based seals are designed for applications where mostly aqueous or buffered-aqueous mobile phases are used. ITB™ graphetized PTFE seals are designed for applications where mobile phases are mostly organic or where chemical compatibility is of primary concern.
  • Description:
    OPTI-MAX EF Inlet Cartridge Check Valve 3/16" Ceramic Ball & Seat, SS Cartridge
  • For use with:
    Waters 225 µL (#3) [510, 590, 600/600E, 610, 650, 6K/6KA]
Accessories
  •  
    Description
  • 83008-150
    Optimize Seal Forming/Insertion Kit, Small/Medium/Large
    Pack of 3 - $344.08
  •  
    Description
  • 83008-152
    Optimize Seal Forming/Insertion Kit, Small
    Each - $145.34
  •  
    Description
  • 83008-154
    Optimize Seal Forming/Insertion Kit, Medium
    Each - $145.34
  •  
    Description
  • 83008-156
    Optimize Seal Forming/Insertion Kit, Large
    Each - $145.34