Orders LinkContactUsLinkComponent
Seals, Optimize Technologies
Seals, Optimize Technologies
OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS Cartridge, 5, 10 & 25 mL S/SC Head, Gilson/Rainin HP, HPX, HPXL, SD200/300, ProStar 210, 215
Catalog # 77100-644
Supplier:   Optimize
Supplier #:  26-56-00638
Seals, Optimize Technologies
Catalog # 77100-644
Supplier:  Optimize
Seals, Optimize Technologies
Seals, Optimize Technologies
OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS Cartridge, 5, 10 & 25 mL S/SC Head, Gilson/Rainin HP, HPX, HPXL, SD200/300, ProStar 210, 215
Price
$687.98
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Piston seals are designed to offer reliable performance in a variety of operating conditions.
  • Polyethylene-based seals are designed for applications where mostly aqueous or buffered-aqueous mobile phases are used
  • Graphetized PTFE seals are designed for applications where mobile phases are mostly organic or where chemical compatibility is of primary concern
  • Reliable performance in a variety of operating conditions
Installing a piston seal can be a tricky undertaking. A relatively small seal must be fit into the seal cavity in such a way to ensure that it is pressed in evenly without distortion or damage. The Optimize Seal Forming/ Insertion Tool Kit was specifically designed to aid in this endeavor. This kit contains three tools to accommodate all of your HPLC/ UHPLC piston seals. One end has a beveled cavity for seal forming, while the other is an insertion support for perfect seal installation. Also included with each kit are complete instructions for maximizing seal life and a solvent compatibility guide to aid in prudent seal material selection.
OPTI-SEAL® UHMW-PE polyethylene-based seals are designed for applications where mostly aqueous or buffered-aqueous mobile phases are used. ITB™ graphetized PTFE seals are designed for applications where mobile phases are mostly organic or where chemical compatibility is of primary concern.
Specifications
  • Description:
    OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS Cartridge, 5, 10 & 25 mL S/SC Head
  • For use with:
    Gilson/Rainin HP, HPX, HPXL, SD200/300, ProStar 210, 215
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Optimize Seal Forming/Insertion Kit, Small/Medium/Large
    Optimize Seal Forming/Insertion Kit, Small/Medium/Large
    83008-150
    Log in for availability
    Pack of 3 - $344.08
Product Information
  • Catalog #
    Availability
    Price
  • Optimize Seal Forming/Insertion Kit, Small
    Optimize Seal Forming/Insertion Kit, Small
    83008-152
    Log in for availability
    Each - $145.34
Product Information
  • Catalog #
    Availability
    Price
  • Optimize Seal Forming/Insertion Kit, Medium
    Optimize Seal Forming/Insertion Kit, Medium
    83008-154
    Log in for availability
    Each - $145.34
Product Information
  • Catalog #
    Availability
    Price
  • Optimize Seal Forming/Insertion Kit, Large
    Optimize Seal Forming/Insertion Kit, Large
    83008-156
    Log in for availability
    Each - $145.34