Seals, Optimize Technologies
Product Details & Documents
Piston seals are designed to offer reliable performance in a variety of operating conditions.
- Polyethylene-based seals are designed for applications where mostly aqueous or buffered-aqueous mobile phases are used
- Graphetized PTFE seals are designed for applications where mobile phases are mostly organic or where chemical compatibility is of primary concern
- Reliable performance in a variety of operating conditions
Installing a piston seal can be a tricky undertaking. A relatively small seal must be fit into the seal cavity in such a way to ensure that it is pressed in evenly without distortion or damage. The Optimize Seal Forming/ Insertion Tool Kit was specifically designed to aid in this endeavor. This kit contains three tools to accommodate all of your HPLC/ UHPLC piston seals. One end has a beveled cavity for seal forming, while the other is an insertion support for perfect seal installation. Also included with each kit are complete instructions for maximizing seal life and a solvent compatibility guide to aid in prudent seal material selection.
OPTI-SEAL® UHMW-PE polyethylene-based seals are designed for applications where mostly aqueous or buffered-aqueous mobile phases are used. ITB™ graphetized PTFE seals are designed for applications where mobile phases are mostly organic or where chemical compatibility is of primary concern.
Specifications
- Description:Piston (Analytical)
- For use with:Bischoff 2200, Anspec SM909, Alcott Micromeritics 760
Accessories
Product Information
- Catalog #AvailabilityPrice
- Optimize Seal Forming/Insertion Kit, Small/Medium/LargeOptimize Seal Forming/Insertion Kit, Small/Medium/Large83008-150Log in for availabilityPack of 3 - $344.08
Product Information
- Catalog #AvailabilityPrice
- Optimize Seal Forming/Insertion Kit, Small
Product Information
- Catalog #AvailabilityPrice
- Optimize Seal Forming/Insertion Kit, Medium
Product Information
- Catalog #AvailabilityPrice
- Optimize Seal Forming/Insertion Kit, Large