Data-Trend Multipurpose Temperature Chart Recorders, Dickson®
7-Day Chart, 0...500 °C/°F
Catalog # 61280-551
Supplier: Dickson
Supplier #: C661
Jump to:
Product Details & Documents
Adaptable to a wide range of applications, these recorders feature a 15.2cm (6") readable chart, AC power with battery backup, user-selectable recording times and temperature ranges, and a remote probe
Increased shielding eliminates electronic noise interference and allows smoother pen movement.
: Recorders include red pen, AC adapter, 9V battery, and 1.2m (4') bead wire probe. Charts must be purchased separately.
: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
Specifications
- Temperature range:0...500 °C/°F
- Description:7-Day Chart
Accessories
Product Information
- Catalog #AvailabilityPrice
- Red Pens