VWR® Talon® Extra-Large Lab-Frames
Lab-Frame with Stainless Steel Rods, Stainless steel
60079-558
Extra-large lab frames offer the greatest versatility for your most demanding setups.
- Extra large design
- For horizontal or vertical use
- Versatile and adaptable
- Mountable on either side
Measure 1220Lx1830Hmm (48x72"). May be used either horizontally or vertically. Include six 305mm (12") rods and feet for securing frame to wall. Apparatus mountable on either side of frame.
: Each Lab Frame includes six 51 mm (2") rods, six 305 mm (12") rods, two 610 mm (24") rods, seven 1220 mm (48") rods, five 1829 mm (72") rods, four rod end connectors, forty-five S-connectors, and eight Lab Frame feet.
- Description:Lab-Frame with Stainless Steel Rods
- Material:Stainless steel