VWR® Talon® Small Lab-Frames
Lab-Frame with Stainless Steel Rods, Stainless steel
Catalog # 60079-552
Supplier: VWR International
Jump to:
Product Details & Documents
Great for glassware setups in small laboratories or where space is limited.
- For vertical mounting
- Ideal for limited-space glassware setups
- Permanently mountable base
Frames measure 610Lx610Hmm (24x24") with a 457mm (18") wide base for stability. Base may be permanently mounted to benchtop with screws (included).
Ordering information: Each Lab Frame includes eight 51 mm (2") rods, two 457 mm (18") rods, eight 610 mm (24") rods, four rod end connectors, eighteen S-connectors, and four Lab Frame feet.
Documents
Specifications
- Description:Lab-Frame with Stainless Steel Rods
- Material:Stainless steel