Orders LinkContactUsLinkComponent
Synthware PTFE Stoppers, Solid and Hollow
Synthware PTFE Stoppers, Solid and Hollow
24/40
Catalog # 60004-906
Supplier:   Kemtech America
Supplier #:  S532440
Synthware PTFE Stoppers, Solid and Hollow
Catalog # 60004-906
Supplier:  Kemtech America
Synthware PTFE Stoppers, Solid and Hollow
Synthware PTFE Stoppers, Solid and Hollow
24/40
Price
$53.74
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
PTFE stopper features a nylon extraction nut that permits easy removal of the stopper from the glass outer joint.
  • Extraction nut
  • Teflon® construction
Available in solid or hollow formats.
Specifications
  • [ST] joint no.:
    24/40