Synthware PTFE Stoppers
24/40
Catalog # 60004-866
Supplier: Kemtech America
: S112440
Jump to:
Product Details & Documents
Stoppers feature PTFE handles for easy gripping.
- T-handle design
- Teflon® construction
Teflon® handle stoppers for easy griping.
Specifications
- [ST] joint no.:24/40