Order Entry
United States
Orders LinkContactUsLinkComponent
Synthware Sure-Grip Separatory Funnels

Synthware Sure-Grip Separatory Funnels

With glass stopper, 1000 ml, 4 mm, 27
Catalog # 60004-250
Supplier:   Kemtech America
Supplier #:  F687000A
Synthware Sure-Grip Separatory Funnels
Catalog # 60004-250
Supplier:  Kemtech America
Synthware Sure-Grip Separatory Funnels
Synthware Sure-Grip Separatory Funnels

With glass stopper, 1000 ml, 4 mm, 27

Price
$235.84
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Globe-shaped separatory funnel features a long stopper neck and a straight section between the funnel body and stopcock that provides a secure grip while shaking

  • Sure-grip design
  • Heavy wall borosilicate construction
  • Ground glass joint

The straight section also aids in clamping. Available with PTFE stopcocks. Funnels are supplied complete with one PTFE or hollow glass stopper.

Specifications
  • Capacity:
    1000 ml
  • Description:
    With glass stopper
  • Stopper no.:
    27
  • Stopcock size:
    4 mm