Synthware Flasks, Erlenmeyer, Threaded, Microscale, Kemtech America
5 ml, 14/10
Catalog # 60004-218
Supplier: Kemtech America
Supplier #: F501005
Synthware Flasks, Erlenmeyer, Threaded, Microscale, Kemtech America
Catalog # 60004-218
Supplier: Kemtech America
Jump to:
Product Details & Documents
Erlenmeyer threaded flask for microscale applications.
- Heavy wall borosilicate construction
- Threaded upper joint
- Open cap included
Supplied with a nylon compression cap, Viton® O-ring, and PTFE-lined silicone septum.
Specifications
- [ST] joint no.:14/10
- Capacity:5 ml
Accessories
Product Information
- Catalog #I.D.×WO-Ring sizeAvailabilityPrice
- Viton® O-Ring, 14/10 threaded joints
Product Information
- Catalog #MaterialCap SizeAvailabilityPrice
- Nylon cap