Orders LinkContactUsLinkComponent
Synthware Distillation Connecting Adapters
Synthware Distillation Connecting Adapters
24/40, [ST] 14/20, 24/40
Catalog # 60000-604
Supplier:   Kemtech America
Supplier #:  A351424
Synthware Distillation Connecting Adapters
Catalog # 60000-604
Supplier:  Kemtech America
Synthware Distillation Connecting Adapters
Synthware Distillation Connecting Adapters
24/40, [ST] 14/20, 24/40
Price
$64.37
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Use distillation connecting adapter as a component in atmospheric or vacuum distallations.
  • Inner joints at bottom and side arm
  • Outer joint at top
The lower end and side arm have [ST] inner joints. Upper end has [ST] outer joint.
Specifications
  • Bottom [ST] joint size:
    24/40
  • Top [ST] joint size:
    [ST] 14/20
  • Side ST joint size:
    24/40