Orders LinkContactUsLinkComponent
Synthware Distillation Connecting Adapters
Synthware Distillation Connecting Adapters
14/20, [ST] 10/18, 14/20
  60000-590
Supplier #:  A351014
Synthware Distillation Connecting Adapters
  60000-590
Synthware Distillation Connecting Adapters
Synthware Distillation Connecting Adapters
14/20, [ST] 10/18, 14/20
$53.47
Each
Minimum order quantity is 1
Use distillation connecting adapter as a component in atmospheric or vacuum distallations.
  • Inner joints at bottom and side arm
  • Outer joint at top
The lower end and side arm have [ST] inner joints. Upper end has [ST] outer joint.
  • Bottom [ST] joint size:
    14/20
  • Top [ST] joint size:
    [ST] 10/18
  • Side ST joint size:
    14/20