VWR® Disposable Tissue Grinders
Product Details & Documents
These disposable tissue grinders are safe and easy to use.
- Disposable
- Safe and easy to use
- Gamma sterilized
Their patented closed system has the ability to grind specimen in a sealed container, helping to prevent contact with the sample
Fully assembled and gamma sterilized. An extra cap and identification label are provided for storage, transport, and added safety. The 35 mL grinder (47732-448) is designed to be freestanding for easier handling.
The molded abrasive surface does not leave any sediment to obstruct examination of the sample.
Documents
Certificates
Search by Lot NumberAvantor Top Rated Guide VWR Histology Brochure.pdf
Product Literature
CoQ VWR Disposable Tissue Grinders.pdf
Conformity declaration
Specifications
- Capacity:50 ml
- Description:Conical