Orders LinkContactUsLinkComponent
EasySat™ Bucketless Floor Mops, Contec®
EasySat™ Bucketless Floor Mops, Contec®
EasySat™ Sterile mop head, 55×46×12 cm (21.5×18.25×4.75"), Knitted Polyester/Foam, 1/bag, 28 bags/case
Catalog # 33501-538
Supplier:   Contec
Supplier #:  BLVKMOPIR
EasySat™ Bucketless Floor Mops, Contec®
Catalog # 33501-538
Supplier:  Contec
EasySat™ Bucketless Floor Mops, Contec®
EasySat™ Bucketless Floor Mops, Contec®
EasySat™ Sterile mop head, 55×46×12 cm (21.5×18.25×4.75"), Knitted Polyester/Foam, 1/bag, 28 bags/case
Price
$868.60
New Customer Promotional Price
Log in for availability
Case of 28
Product Details & Documents
Designed specifically for cleanroom use, this mop system completely eliminates the need for bucket or wringer.
  • Hydrophilic polyester/clean ester, laser cut sealed edge foam material
  • Translucent tank allows user to see solution
  • Fully autoclavable for critical areas
  • Easy to use
  • Compatible with a wide range of solvents
System includes a 1.5 l (52 oz.) translucent solution tank that can clean up to 5,000 square feet. Tank weighs only 3.2 kg (7 lbs.) when full. A push button flow control releases cleaning solution on demand. The angled, 55.9 cm (22") mop heads clean hard-to-reach areas and can be attached and removed within seconds.
System and mop heads are fully autoclavable up to 121 °C (250 °F) for 30 minutes. Mop heads come in regular and textured styles and are also available gamma irradiated.
It features lasercut sealed-edge VertiKlean® mop head material, which is a lightweight polyester fabric laminated to clean ester foam for excellent abrasion resistance.
 : Mop system and mop heads must be purchased separately.
Specifications
  • Description:
    EasySat™ Sterile mop head
  • Dimensions:
    55×46×12 cm (21.5×18.25×4.75")
  • Material:
    Knitted Polyester/Foam
  • Packed:
    1/bag, 28 bags/case