Order Entry
United States
Orders LinkContactUsLinkComponent
Operating Scissors, Integra™ Miltex®

Operating Scissors, Integra™ Miltex®

Sharp/Blunt, 15.2 cm (6"), Straight
Catalog # 21909-286
Supplier:   Miltex
Supplier #:  5-17
Operating Scissors, Integra™ Miltex®
Catalog # 21909-286
Supplier:  Miltex
Operating Scissors, Integra™ Miltex®
Operating Scissors, Integra™ Miltex®

Sharp/Blunt, 15.2 cm (6"), Straight

Price
$114.23
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Standard pattern

Stainless steel

Specifications
  • Description:
    Sharp/Blunt
  • Length:
    15.2 cm (6")
  • Blade type:
    Straight

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.