700 System Premium Receiver Head Assemblies, Techcon
1.8 m (6")
Catalog # 14219-724
Supplier: Techcon Systems
Supplier #: 71006RHB
Jump to:
Product Details & Documents
Head assemblies ensure accident-proof dispensing and provide a secure and reliable link between the dispensing barrel and the controller
Complete dispensing control. Compatible with all industry-standard dispensing barrels. Designed to exact industry standards.
Specifications
- Tubing length:1.8 m (6")