SE Series Dispensing Tips, Techcon
Green, 18
Catalog # 14219-600
Supplier: Techcon Systems
Supplier #: SE418050PK
Jump to:
Product Details & Documents
Tips have a polypropylene hub and stainless steel cannula
Tips are 1.27cm (1/2") long. Colors indicate gauge size.
Specifications
- Color:Green
- Gauge:18