VWR® Modular Heating Blocks, Solid
Solid Block, Single, Single, 9.5 x 7.6 x 5.1 cm (33/4 x 3 x 2")
13259-288
Constructed from a solid anodized aluminum block.
- Now with Best-in-Class, Exclusive 5-Year Warranty
For use as a low-temperature hotplate or for drilling a customized block. Available in single or double block size.
- Description:Solid Block, Single
- L×W×H:9.5 x 7.6 x 5.1 cm (33/4 x 3 x 2")
- Block size:Single



