Orders LinkContactUsLinkComponent
Dissolution Sampling Cannula, Quality Lab Accessories
Dissolution Sampling Cannula, Quality Lab Accessories
4.75’’ (120 mm) Bent 316 SS Cannula with Stainless Steel Luer Lock and Perm Stainless steel Tip for QLA "01" style filters
Catalog # 11010-092
Supplier:   QLA
Supplier #:  CAN475-VK
Dissolution Sampling Cannula, Quality Lab Accessories
Catalog # 11010-092
Supplier:  QLA
Dissolution Sampling Cannula, Quality Lab Accessories
Dissolution Sampling Cannula, Quality Lab Accessories
4.75’’ (120 mm) Bent 316 SS Cannula with Stainless Steel Luer Lock and Perm Stainless steel Tip for QLA "01" style filters
Price
$235.59
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
QLA manufactures a wide variety of Sampling Cannula and Probes for the Dissolution Industry.
  • Manual sampling cannula are manufactured in electroplished 316 Stainless Steel or PEEK materials and come in a variety of lengths
Many industry standard cannula configurations and unique, proprietary QLA configurations are available for most manual and automated sampling applications
The USP defines the sampling point as: "A zone half way between the top of the media and top of the paddle or basket not less than 1 cm from the vessel wall. The sampling position is critical but so too is the type of sampling cannula (also known as a probe) used."
Optional stoppers set the proper sampling position. Manual cannula configurations accommodate all standard tip filters.
Specifications
  • Description:
    4.75’’ (120 mm) Bent 316 SS Cannula with Stainless Steel Luer Lock and Perm Stainless steel Tip for QLA "01" style filters