Orders LinkContactUsLinkComponent
Dissolution Sampling Cannula, Quality Lab Accessories
4.75" (120 mm) Bent PEEK 1/8" (3.2 mm) OD Cannula with PEEK Luer Lock and PTFE sleeve for QLA "01" style filters
Catalog # 11010-054
Supplier:   QLA
 :  CA8475-PV
Dissolution Sampling Cannula, Quality Lab Accessories
Catalog # 11010-054
Supplier:  QLA
Dissolution Sampling Cannula, Quality Lab Accessories
Dissolution Sampling Cannula, Quality Lab Accessories
4.75" (120 mm) Bent PEEK 1/8" (3.2 mm) OD Cannula with PEEK Luer Lock and PTFE sleeve for QLA "01" style filters
Price
$86.84
Log in for availability
Each
Product Details & Documents
QLA manufactures a wide variety of Sampling Cannula and Probes for the Dissolution Industry.
  • Manual sampling cannula are manufactured in electroplished 316 Stainless Steel or PEEK materials and come in a variety of lengths
Many industry standard cannula configurations and unique, proprietary QLA configurations are available for most manual and automated sampling applications
The USP defines the sampling point as: "A zone half way between the top of the media and top of the paddle or basket not less than 1 cm from the vessel wall. The sampling position is critical but so too is the type of sampling cannula (also known as a probe) used."
Optional stoppers set the proper sampling position. Manual cannula configurations accommodate all standard tip filters.
Specifications
  • Description:
    4.75" (120 mm) Bent PEEK 1/8" (3.2 mm) OD Cannula with PEEK Luer Lock and PTFE sleeve for QLA "01" style filters