Order Entry
United States
Orders LinkContactUsLinkComponent
solution 100 mg/mL

solution 100 mg/mL

1 ml
Catalog # 101449-080
solution 100 mg/mL
Catalog # 101449-080
Supplier:  
CAS Number:  4697-36-3
solution 100 mg/mL

1 ml

CAS Number:  4697-36-3
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
CARBENICILLIN SOLUTION 100MG Catalog #77443-474

Product 77443-474 is a replacement for 101449-080 but varies by Brand, Packaging or Quantity.

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Carbenicillin is a semi-synthetic ampicillin analog, and like ampicillin is a beta-lactam antibiotic. Carbenicillin is a member of the carboxypenicillin subgroup of the penicillins.

It is effective against Gram-negative organisms including H. influenzae and E. coli. Carbenicillin is also used in transformed Agrobacter species. Carbenicillin was the first penicillin with significant activity against Pseudomonas species.

  • CAS Number:4697-36-3
  • Formula:C₁₇H₁₈N₂O₆S

Documents

Specifications

Specifications

  • Size:
    1 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.