Order Entry
United States
Orders LinkContactUsLinkComponent
BUFFER_PBS_PH-7.4_6X1000ML

BUFFER_PBS_PH-7.4_6X1000ML

Catalog # MSPP-10010072
Marketsource
Supplier:   Invitrogen
Supplier #:  10010072
BUFFER_PBS_PH-7.4_6X1000ML
Catalog # MSPP-10010072
Supplier:  Invitrogen
Marketsource
CAS Number:  
BUFFER_PBS_PH-7.4_6X1000ML
CAS Number:  
Price
$438.46
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.