Thread Adapters, Polypropylene
External thread: S 70×6, Internal thread: G 3/4", White
Catalog # 78649-720
New Product
Supplier: BURKLE INC
Supplier #: 0600-0130
Jump to:
Product Details & Documents
Bürkle offers a broad range of thread adapters to facilitate secure connections between many Bürkle pumps and discharge cocks to a variety of containers
This wide variety of adapters ensures users can find the correct connection.
Specifications
- Color:White
- Description:External thread: S 70×6, Internal thread: G 3/4"