Order Entry
United States
Orders LinkContactUsLinkComponent
D-(+)-Mannose 99%

D-(+)-Mannose 99%

Bottle, 500 g
Catalog # MPAAA10842-36
New Product
Supplier #:  A10842-36
D-(+)-Mannose 99%
Catalog # MPAAA10842-36
New Product
CAS Number:  3458-28-4
D-(+)-Mannose 99%
D-(+)-Mannose 99%

Bottle, 500 g

CAS Number:  3458-28-4
Price
$370.50
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

MDL: MFCD00064122 Beilstein Registry No.: 1564373 Soluble in water Optical Rotation: +13.8°C (c=10 in water, 24h)

Purity: 99% CAS No.: 3458-28-4 Molecular Formula: C6H12O6 Formula Weight: 180.16 Melting Point: 127-133° Solubility: Soluble in water Beilstein Reference: 1564373 Merck Reference: 14,5747 Optical Rotation: +13.8° (c=10 in water, 24h) MDL No.: MFCD00064122

Synonyms:  (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
  • CAS Number:3458-28-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00064122
  • Molecular Weight:180.16 g/mol
  • Merck Index:14,5747
  • Boiling Pt:376 °C (1013 hPa)
  • Melting Pt:127…133 °C
  • Density:1.54
  • Storage Temperature:Ambient
  • EINECS:222-392-4
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.