Mining Sampling Bags, Clear Poly
Poly sample bag, 4 mil, 10×16", Plastic, Clear
80701-994
New Product
: 1016390500CL
Clear poly, mining sampling bags in various sizes and mil thicknesses.
Suitable for field and lab applications
- Made in Canada
Side welded seal imparts improved, with greater strength, and a resulting solid tubing bottom edge.
- Color:Clear
- Description:Poly sample bag
- Dimensions:10×16"
- Material:Plastic
- Thickness:4 mil