Orders LinkContactUsLinkComponent
Ward's® Variable Standing Wave Generator
Ward's® Variable Standing Wave Generator
Variable standing wave generator
Catalog # 470360-230
New Product
Supplier:   VWR International
Ward's® Variable Standing Wave Generator
Catalog # 470360-230
Supplier:  VWR International
New Product
Ward's® Variable Standing Wave Generator
Ward's® Variable Standing Wave Generator
Variable standing wave generator
Price
$39.60
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
An exciting, interactive exploration, this generator includes various mass attachments to investigate degrees of string tension in standing waves.
  • Ø×L: 3×15 cm
  • String length: 5.5 m
  • Weight: 0.5 kg
The device features a battery-operated motor with an off balanced bolt attached to the shaft. When running, this motor creates a vibration in the string that sets up a standing wave along the entire length of string. The nuts on the bottom alow you to change the center of mass, and therefore change the frequency of the standing waves. The top eyebolt allows slotted masses to be attached, so the affect of string tension on the wavelength can also be investigated. Developed by Bruce Yeany.
Specifications
  • Description:
    Variable standing wave generator