Orders LinkContactUsLinkComponent
Milwaukee® Basic Waterproof EC Testers
Milwaukee® Basic Waterproof EC Testers
Replacement probe C66
Catalog # 470359-670
New Product
Supplier #:  MA73076
Milwaukee® Basic Waterproof EC Testers
Catalog # 470359-670
New Product
Milwaukee® Basic Waterproof EC Testers
Milwaukee® Basic Waterproof EC Testers
Replacement probe C66
Price
$32.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
These durable EC testers feature a replaceable electrode and are ideal for student use in the classroom and field environments.
  • EC testers that are fast and easy to read with simple one-point calibration
  • Two-year warranty (six months on probe)
  • Automatic temperature compensation (ATC)
  • Over 250 hours of continuous battery life (batteries included)
These models combine a rugged waterproof construction with a replaceable electrode - offering advanced EC accuracy and long term value that outperforms basic testers, making it ideal for repeated use in K-12 science labs and field work.
Ordering information: Ordering additional calibration solution is recommended.
Delivery information: Batteries, probe, and calibration starter pack included.
Specifications
  • Description:
    Replacement probe C66