Milwaukee® Basic Waterproof EC Testers
These durable EC testers feature a replaceable electrode and are ideal for student use in the classroom and field environments.
EC testers that are fast and easy to read with simple one-point calibration
- Two-year warranty (six months on probe)
- Automatic temperature compensation (ATC)
- Over 250 hours of continuous battery life (batteries included)
These models combine a rugged waterproof construction with a replaceable electrode - offering advanced EC accuracy and long term value that outperforms basic testers, making it ideal for repeated use in K-12 science labs and field work.
: Ordering additional calibration solution is recommended.
: Batteries, probe, and calibration starter pack included.
- Model:C66
- Description:High range EC tester
- Cat. No.:470359-570
- Conductivity Range:0.00 - 10.00 mS/cm
- Conductivity Accuracy:±2% Full scale