Milwaukee® Basic Waterproof EC Testers
Low range EC tester, C65
Catalog # 470359-568
New Product
Supplier: MILWAUKEE INSTRUMENTS, INC.
Supplier #: C65
Milwaukee® Basic Waterproof EC Testers
Catalog # 470359-568
Supplier: MILWAUKEE INSTRUMENTS, INC.
New Product
Jump to:
Product Details & Documents
These durable EC testers feature a replaceable electrode and are ideal for student use in the classroom and field environments.
EC testers that are fast and easy to read with simple one-point calibration
- Two-year warranty (six months on probe)
- Automatic temperature compensation (ATC)
- Over 250 hours of continuous battery life (batteries included)
These models combine a rugged waterproof construction with a replaceable electrode - offering advanced EC accuracy and long term value that outperforms basic testers, making it ideal for repeated use in K-12 science labs and field work.
Ordering information: Ordering additional calibration solution is recommended.
Delivery information: Batteries, probe, and calibration starter pack included.
Documents
Specifications
- Model:C65
- Description:Low range EC tester
- Cat. No.:470359-568
- Conductivity Range:0 - 1999 uS/cm
- Conductivity Accuracy:±2% Full scale