Orders LinkContactUsLinkComponent
Bis(2-methoxyethyl)ether ≥99% stabilised
Glass bottle, 500 g
  78067-824
New Product
 :   Ambeed
 :  A449960-500G
Bis(2-methoxyethyl)ether ≥99% stabilised
  78067-824
 :  Ambeed
New Product
 :  111-96-6
Bis(2-methoxyethyl)ether ≥99% stabilised
Glass bottle, 500 g
 :  111-96-6
$66.00
Each(1/Itm/Bx)
 : for Research Use Only
 :  2-Methoxyethyl ether, 1-Methoxy-2-(2-methoxyethoxy)ethane, Diethylene glycol dimethyl ether, Dimethyldigol, Diglyme
  •  :111-96-6
  •  :C₆H₁₄O₃
  •  :MFCD00008503
  •  :134.18 g/mol
  •  :3,Ⅲ
  •  :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 0.01%
  •  :3271
  • Pack type:
    Glass bottle
  • Size:
    500 g