Order Entry
United States
Orders LinkContactUsLinkComponent
VWR® Multi-Function Liquid Transfer Cap System

VWR® Multi-Function Liquid Transfer Cap System

Multi function transfer cap, 5 L shaker flask
Catalog # 76404-502
Supplier:   VWR International
VWR® Multi-Function Liquid Transfer Cap System
Catalog # 76404-502
Supplier:  VWR International
VWR® Multi-Function Liquid Transfer Cap System

Multi function transfer cap, 5 L shaker flask

Price
$1,508.04
New Customer Promotional Price
Log in for availability
Case of 4
Product Details & Documents

Multi-function liquid transfer cap system for 5 L Erlenmeyer flasks, sterile.

  • Closed system reduces the risks of contamination during liquid transfer

  • The cap is moulded and connected in an integrated manner, reducing the risks of leakage and media residues
  • High-quality materials and smooth inner wall of the tubing provide an excellent transfer performance
  • Sterility level (SAL) is maintained at 10⁻⁶
  • Endotoxin-free, and no ingredients of animal origin

Unlike the two-way liquid transfer cap, the multifunctional transfer cap can be directly placed in an incubator for culture after the liquid transfer is completed. It can reach a large air flux. The sampling part is composed of a sampling nozzle and a one-way valve, which can prevent the liquid from flowing backwards during the sampling process and ensure the aseptic sampling. The liquid inlet tubing is provided with a PTFE needle filter, which solves the issue of liquid remaining in the tubing during the feeding process.

Certifications: In compliance with IS0 11137. Samples validated based on Tachypleus Amebocyte Lysate test. Tested for the attribute of Cell Attachment and Growth.

Specifications
  • Description:
    Multi function transfer cap
  • For use with:
    5 L shaker flask
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.