Precision Scissors, 4¹/₂"
4¹/₂" precision scissors, 1³/₈", 0.05 lbs.
Catalog # 78621-282
New Product
Supplier: Aven Tools
Supplier #: 11016
Jump to:
Product Details & Documents
Fine beaks for small places, precise and delicate work.
- Fully polished stainless steel provides excellent corrosion resistance
- Durable stainless steel blade
Specifications
- Description:4¹/₂" precision scissors
- Blade length:1³/₈"
- Weight:0.05 lbs.