Orders LinkContactUsLinkComponent
Recirculating Chillers, VALEGRO
Recirculating Chillers, VALEGRO
New Product
Supplier:   JULABO
Recirculating Chillers, VALEGRO
Supplier:  JULABO
New Product
Certificates
Recirculating Chillers, VALEGRO
Recirculating Chillers, VALEGRO
23 Options Available Below
Product Details & Documents
VALEGRO natural refrigerant recirculating chillers from JULABO provide an environmentally friendly option for a wide range of cooling and heating tasks.
  • Climate-friendly natural refrigerant
    OLED touch display with intuitive menu navigation
    USB-C and RS232 I/O, Ethernet available as an option
    Illuminated, easy-to-clean level indicator
    Adjustable pump to regulate pressure and flow rate
    'H' units with integrated heater to 80 °C
VALEGRO chillers have an operating range of −20 to +40 °C, ±0.3 °C (except for VALEGRO 250, −10 to +40 °C) and 'H' models include a heater for an operating range of 20 to 80 °C, ±0.2 °C. The OLED touch screen interface makes operation and menu access easy. An adjustable pump enables operation to the users flow/pressure needs.
Integrated timer and autostart functions. RS232 and USB-C interfaces facilitate external control via PC with optional Ethernet activation (requires 30004147). Data recording via the USB-C port for time/temperature data and Blackbox data capture for remote service diagnosis. All interfaces are located directly under the front display. Illuminated fluid level indicator (except on VALEGRO 250) includes a brush for easy cleaning. For use with water and water/glycol bath fluids.
Easy maintenance with removable front grid (except for the VALEGRO 250). We recommend periodic inspection of the condenser - vacuum away any accumulated dust/debris.
Max. flow rate/max. pressure:
23 L/min, 16 PSI
VALEGRO 1203/1203H, 1503/1503 H, 1803/1803H, 2503/2503H: 40 L/min, 50.8 PSI
Certifications: IP protection class according to DIN EN 60529: IP 21
All models have CE certification.
Ordering information: Accessories such as bath fluid, tubing, clamps must be ordered separately.
Delivery information: VALEGRO chillers with 1.1 bar | 16 PSI with M16×1 threaded pump connections with 8 and 12 mm hose barbs; Chillers with 3.5 bar (50.8 PSI) pump with M24×1.5 threaded pump connections and 12 mm hose barbs. All units carry a 2-year return-to-factory warranty starting from shipment date.
Caution: Contains pressurized hydrocarbon refrigerant. Read the operating manual. Install in area that meets the minimum space requirements.   Cooling capacity at +20; 0: −10 and −20 °C.
Product Family Options
Models with 100 - 115 V
Product Information
  • Model
    Temperature range
    Electrical
    W×D×H
    Cooling capacity
    Fill volume
    Pump flow rate
    Pump pressure
    Heating power
    Availability
    Price
  • VALEGRO 1001H
    −20...+40 °C
    200 − 230 V, 50/60 Hz
    Ext: 36.6×52.6×61 cm (14.4×20.7×24")
    1800; 1300; 800; 400 W
    4 - 7 L
    23 L/min
    16 PSI
    1 kW
    Log in for availability
    Log in for availability
    Each
    Each - $16,588.00
    $16,588.00
    Each
       $16,588.00
    Minimum order quantity is 1
    Supplier Number: 9612101.3000566196
    Specifications:
    New Product
    New Product
  • VALEGRO 801H
    −20...+80 °C
    200 − 230 V, 50/60 Hz
    Ext: 36.6×52.6×61 cm (14.4×20.7×24")
    1200; 900; 600; 350 W
    4 - 7 L
    23 L/min
    16 PSI
    1 kW
    Log in for availability
    Log in for availability
    Each
    Each - $15,312.00
    $15,312.00
    Each
       $15,312.00
    Minimum order quantity is 1
    Supplier Number: 9612081.3000565097
    Specifications:
    New Product
    New Product
  • VALEGRO 1201H
    −20...+80 °C
    200 − 230 V, 50/60 Hz
    Ext: 36.6×52.6×61 cm (14.4×20.7×24")
    1500; 1000; 700; 400 W
    4 - 7 L
    23 L/min
    16 PSI
    1 kW
    Log in for availability
    Log in for availability
    Each
    Each - $15,216.00
    $15,216.00
    Each
       $15,216.00
    Minimum order quantity is 1
    Supplier Number: 9612121.3000568096
    Specifications:
    New Product
    New Product
  • VALEGRO 1501H
    −20...+80 °C
    200 − 230 V, 50/60 Hz
    Ext: 36.6×52.6×61 cm (14.4×20.7×24")
    1800; 1100; 750; 470 W
    4 - 7 L
    23 L/min
    16 PSI
    1 kW
    Log in for availability
    Log in for availability
    Each
    Each - $19,155.00
    $19,155.00
    Each
       $19,155.00
    Minimum order quantity is 1
    Supplier Number: 9612151.3000568207
    Specifications:
    New Product
    New Product
  • VALEGRO 1801H
    −20...+80 °C
    200 − 230 V, 50/60 Hz
    Ext: 36.6×52.6×61 cm (14.4×20.7×24")
    2503; 1450; 1000; 500 W
    4 - 7 L
    23 L/min
    16 PSI
    1 kW
    Log in for availability
    Log in for availability
    Each
    Each - $19,801.00
    $19,801.00
    Each
       $19,801.00
    Minimum order quantity is 1
    Supplier Number: 9612181.3000569195
    Specifications:
    New Product
    New Product
Models with 100 - 230 V
Product Information
  • Model
    Temperature range
    Electrical
    W×D×H
    Cooling capacity
    Fill volume
    Pump flow rate
    Pump pressure
    Heating power
    Availability
    Price
  • VALEGRO 801
    −20...+40 °C
    100 − 230 V, 50/60 Hz
    Ext: 36.6×52.6×61 cm (14.4×20.7×24")
    800; 650; 450; 250 W
    4 - 7 L
    23 L/min
    16 PSI
    Log in for availability
    Log in for availability
    Each
    Each - $11,994.00
    $11,994.00
    Each
       $11,994.00
    Minimum order quantity is 1
    Supplier Number: 9610081.3000560999
    Specifications:
    New Product
    New Product
  • VALEGRO 1001
    −20...+40 °C
    100 − 230 V, 50/60 Hz
    Ext: 36.6×52.6×61 cm (14.4×20.7×24")
    1000; 750; 500; 280 W
    4 - 7 L
    23 L/min
    16 PSI
    Log in for availability
    Log in for availability
    Each
    Each - $13,015.00
    $13,015.00
    Each
       $13,015.00
    Minimum order quantity is 1
    Supplier Number: 9610101.30005618
    Specifications:
    New Product
    New Product
  • VALEGRO 1201
    −20...+40 °C
    100 − 230 V, 50/60 Hz
    Ext: 36.6×52.6×61 cm (14.4×20.7×24")
    1200; 900; 600; 350 W
    4 - 7 L
    23 L/min
    16 PSI
    Log in for availability
    Log in for availability
    Each
    Each - $10,758.00
    $10,758.00
    Each
       $10,758.00
    Minimum order quantity is 1
    Supplier Number: 9610121.3000562694
    Specifications:
    New Product
    New Product
  • VALEGRO 1501
    −20...+40 °C
    100 − 230 V, 50/60 Hz
    Ext: 36.6×52.6×61 cm (14.4×20.7×24")
    1500; 1000; 700; 400 W
    4 - 7 L
    23 L/min
    16 PSI
    Log in for availability
    Log in for availability
    Each
    Each - $14,420.00
    $14,420.00
    Each
       $14,420.00
    Minimum order quantity is 1
    Supplier Number: 9610151.3000563495
    Specifications:
    New Product
    New Product
  • VALEGRO 1801
    −20...+80 °C
    200 − 230 V, 50/60 Hz
    Ext: 36.6×52.6×61 cm (14.4×20.7×24")
    800; 650; 450; 250 W
    4 - 7 L
    23 L/min
    16 PSI
    Log in for availability
    Log in for availability
    Each
    Each - $15,497.00
    $15,497.00
    Each
       $15,497.00
    Minimum order quantity is 1
    Supplier Number: 9610181.3000564296
    Specifications:
    New Product
    New Product
Models with 200 - 230 V
Product Information
  • Model
    Temperature range
    Electrical
    W×D×H
    Cooling capacity
    Fill volume
    Pump flow rate
    Pump pressure
    Heating power
    Availability
    Price
  • VALEGRO 1203
    −20...+40 °C
    200 − 230 V, 50/60 Hz
    Ext: 42.9×50.0×69.1 cm (16.9×19.7×27.2")
    1200; 900; 500; 300 W
    3 - 8 L
    40 L/min
    50.8 PSI
    Log in for availability
    Log in for availability
    Each
    Each - $16,126.00
    $16,126.00
    Each
       $16,126.00
    Minimum order quantity is 1
    Supplier Number: 9610123.3000697903
    Specifications:
    New Product
    New Product
  • VALEGRO 1503
    −20...+40 °C
    200 − 230 V, 50/60 Hz
    Ext: 42.9×50.0×69.1 cm (16.9×19.7×27.2")
    1500; 1000; 700; 350 W
    3 - 8 L
    40 L/min
    50.8 PSI
    Log in for availability
    Log in for availability
    Each
    Each - $16,567.00
    $16,567.00
    Each
       $16,567.00
    Minimum order quantity is 1
    Supplier Number: 9610153.3000698499
    Specifications:
    New Product
    New Product
  • VALEGRO 801H
    −20...+40 °C
    100 − 230 V, 50/60 Hz
    Ext: 36.6×52.6×61 cm (14.4×20.7×24")
    1800; 1100; 750; 470 W
    4 - 7 L
    23 L/min
    16 PSI
    1.5 - 2 kW
    Log in for availability
    Log in for availability
    Each
    Each - $15,312.00
    $15,312.00
    Each
       $15,312.00
    Minimum order quantity is 1
    Supplier Number: 9612081.3000565302
    Specifications:
    New Product
    New Product
  • VALEGRO 1001H
    −20...+40 °C
    200 − 230 V, 50/60 Hz
    Ext: 36.6×52.6×61 cm (14.4×20.7×24")
    2500; 1450; 1000; 500 W
    4 - 7 L
    23 L/min
    16 PSI
    1.5 - 2 kW
    Log in for availability
    Log in for availability
    Each
    Each - $16,588.00
    $16,588.00
    Each
       $16,588.00
    Minimum order quantity is 1
    Supplier Number: 9612101.3000566307
    Specifications:
    New Product
    New Product
  • VALEGRO 1201H
    −20...+80 °C
    100 − 115 V, 50/60 Hz
    Ext: 36.6×52.6×61 cm (14.4×20.7×24")
    800; 650; 450; 250 W
    4 - 7 L
    23 L/min
    16 PSI
    1.5 - 2 kW
    Log in for availability
    Log in for availability
    Each
    Each - $15,216.00
    $15,216.00
    Each
       $15,216.00
    Minimum order quantity is 1
    Supplier Number: 9612121.3000567202
    Specifications:
    New Product
    New Product
  • VALEGRO 1803
    −20...+80 °C
    100 − 115 V, 50/60 Hz
    Ext: 42.9×50.0×69.1 cm (16.9×19.7×27.2")
    1000; 750; 500; 280 W
    3 - 8 L
    40 L/min
    50.8 PSI
    Log in for availability
    Log in for availability
    Each
    Each - $17,450.00
    $17,450.00
    Each
       $17,450.00
    Minimum order quantity is 1
    Supplier Number: 9610183.3000699095
    Specifications:
    New Product
    New Product
  • VALEGRO 2503
    −20...+80 °C
    200 − 230 V, 50/60 Hz
    Ext: 42.9×50.0×69.1 cm (16.9×19.7×27.2")
    1000; 750; 500; 280 W
    3 - 8 L
    40 L/min
    50.8 PSI
    Log in for availability
    Log in for availability
    Each
    Each - $17,226.00
    $17,226.00
    Each
       $17,226.00
    Minimum order quantity is 1
    Supplier Number: 9610253.3000699803
    Specifications:
    New Product
    New Product
  • VALEGRO 1801H
    −20...+80 °C
    100 − 115 V, 50/60 Hz
    Ext: 36.6×52.6×61 cm (14.4×20.7×24")
    1200; 900; 600; 350 W
    4 - 7 L
    23 L/min
    16 PSI
    1 kW
    Log in for availability
    Log in for availability
    Each
    Each - $19,801.00
    $19,801.00
    Each
       $19,801.00
    Minimum order quantity is 1
    Supplier Number: 9612181.3000569306
    Specifications:
    New Product
    New Product
  • VALEGRO 1803H
    −20...+80 °C
    200 − 230 V, 50/60 Hz
    Ext: 42.9×50.0×69.1 cm (16.9×19.7×27.2")
    1200; 900; 500; 300 W
    3 - 8 L
    40 L/min
    50.8 PSI
    1.5 - 2 kW
    Log in for availability
    Log in for availability
    Each
    Each - $21,868.00
    $21,868.00
    Each
       $21,868.00
    Minimum order quantity is 1
    Supplier Number: 9612183.3000701703
    Specifications:
    New Product
    New Product
  • VALEGRO 2503H
    −20...+80 °C
    100 − 115 V, 50/60 Hz
    Ext: 42.9×50.0×69.1 cm (16.9×19.7×27.2")
    1500; 1000; 700; 400 W
    3 - 8 L
    40 L/min
    50.8 PSI
    1.5 - 2 kW
    Log in for availability
    Log in for availability
    Each
    Each - $23,867.00
    $23,867.00
    Each
       $23,867.00
    Minimum order quantity is 1
    Supplier Number: 9612253.3000702299
    Specifications:
    New Product
    New Product
  • VALEGRO 1203H
    −20...+80 °C
    200 − 230 V, 50/60 Hz
    Ext: 42.9×50.0×69.1 cm (16.9×19.7×27.2")
    1500; 1000; 700; 350 W
    3 - 8 L
    40 L/min
    50.8 PSI
    1.5 - 2 kW
    Log in for availability
    Log in for availability
    Each
    Each - $20,101.00
    $20,101.00
    Each
       $20,101.00
    Minimum order quantity is 1
    Supplier Number: 9612123.3000700399
    Specifications:
    New Product
    New Product
  • VALEGRO 1501H
    −20...+80 °C
    100 − 115 V, 50/60 Hz
    Ext: 36.6×52.6×61 cm (14.4×20.7×24")
    1800; 1100; 750; 470 W
    4 - 7 L
    23 L/min
    16 PSI
    1 kW
    Log in for availability
    Log in for availability
    Each
    Each - $19,155.00
    $19,155.00
    Each
       $19,155.00
    Minimum order quantity is 1
    Supplier Number: 9612151.30005683
    Specifications:
    New Product
    New Product
  • VALEGRO 1503H
    −20...+80 °C
    200 − 230 V, 50/60 Hz
    Ext: 42.9×50.0×69.1 cm (16.9×19.7×27.2")
    1800; 1300; 800; 400 W
    3 - 8 L
    40 L/min
    50.8 PSI
    1.5 - 2 kW
    Log in for availability
    Log in for availability
    Each
    Each - $20,543.00
    $20,543.00
    Each
       $20,543.00
    Minimum order quantity is 1
    Supplier Number: 9612153.3000701107
    Specifications:
    New Product