CellPro™ Normal Goat Serum, US Source
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Normal Goat Serum, US source is a high-quality biological reagent collected from non-immunized goats and commonly used in immunology and diagnostic applications. It is widely utilized as a blocking reagent, negative control, or diluent in antibody-based assays to reduce non-specific binding. US-source collection ensures traceability, consistency, and reliable performance across research workflows.
- Collected from non-immunized US-source goats
- Ideal for blocking and reducing non-specific binding
- Commonly used as a negative control or antibody diluent
- Suitable for ELISA, immunohistochemistry, and immunoassays
- Consistent protein composition and performance
- US source for enhanced traceability and quality assurance
Normal Goat Serum, US Source is a premium biological reagent derived from non-immunized goats for use in immunological research and diagnostic assay development. Rich in serum proteins but free from specific induced antibodies, it is commonly used as a blocking agent to minimize non-specific binding, as a negative control, or as a diluent in antibody-based applications such as ELISA, immunohistochemistry (IHC), and other immunoassays.
Sourced from healthy goats in the United States, this serum offers dependable traceability, lot-to-lot consistency, and reliable assay performance. Each lot is quality tested to ensure suitability for laboratory and research use, making it a trusted component in routine and advanced immunological workflows.
Specifications
- Serum format:Frozen
- Sterility:Sterile
- Description:Normal goat serum, US source
- Size:500 ml
- Packaging:Bottle
- Cat. No.:MSPP-SF21ML-B
- Storage Temperature:–20 °C or colder
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



