Orders LinkContactUsLinkComponent
Maestro Non-Elevating Leg Frames
Maestro Non-Elevating Leg Frames
New Product
Supplier:   MOPEC
Maestro Non-Elevating Leg Frames
Supplier:  MOPEC
New Product
Certificates
Maestro Non-Elevating Leg Frames
Maestro Non-Elevating Leg Frames
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Designed to be configured for specific laboratory requirements, the Maestro Encore provides a platform of optional accessories to improve workspace ergonomics and productivity.
  • 304 stainless steel material, 180 grit finish
  • Compatible with all Maestro and Maestro Encore Non-Elevating grossing stations
Designed for customers who do not need the elevation function on a standard Elevating Maestro or Maestro Encore grossing station. To add to an existing Countertop Maestro or Maestro Encore grossing station, please contact a sales representative.
Delivery information: Includes leg frame and leveling feet with 1.625" of adjustment, 11 gal 304 stainless steel seismic anchor brackets included, with ³/₈" 304 stainless hardware.
Product Family Options
Product Information
  • Material
    Ext. dimensions
    Availability
    Price
  • 78142-292
    Maestro non-elevating leg frame
    Maestro non-elevating leg frame
    Stainless steel
    48L×29W×32" H
    Log in for availability
    Log in for availability
    Each
    Each - $1,596.20
    $1,596.20
    Each
       $1,596.20
    Supplier Number: MB1030-048
    Specifications:
    New Product
    New Product
  • 78142-293
    Maestro non-elevating leg frame
    Maestro non-elevating leg frame
    Stainless steel
    60L×29W×32" H
    Log in for availability
    Log in for availability
    Each
    Each - $1,596.20
    $1,596.20
    Each
       $1,596.20
    Supplier Number: MB1030-060
    Specifications:
    New Product
    New Product
  • 78142-294
    Maestro non-elevating leg frame
    Maestro non-elevating leg frame
    Stainless steel
    72L×29W×32" H
    Log in for availability
    Log in for availability
    Each
    Each - $1,596.20
    $1,596.20
    Each
       $1,596.20
    Supplier Number: MB1030-072
    Specifications:
    New Product
    New Product
  • 78142-295
    Maestro non-elevating leg frame
    Maestro non-elevating leg frame
    Stainless steel
    96L×29W×32" H
    Log in for availability
    Log in for availability
    Each
    Each - $1,596.20
    $1,596.20
    Each
       $1,596.20
    Supplier Number: MB1030-096
    Specifications:
    New Product