Orders LinkContactUsLinkComponent
Control Slides
Control Slides
Control slides, IHC, Treponema pallidum, 25/pack
Catalog # 76947-810
Supplier:   Mopec
Supplier #:  SC619-25
Control Slides
Catalog # 76947-810
Supplier:  Mopec
Control Slides
Control Slides
Control slides, IHC, Treponema pallidum, 25/pack
Price
$815.39
Log in for availability
Case of 25
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Prepared with precision to ensure consistency and reliability, each box contains 25 slides, ideal for use in histology and pathology laboratories.
  • Box of 25 contains 24 unstained slides and one stained slide
Expertly prepared for consistency and quality, these control slides provide reliable reference material for histology and pathology applications.
Specifications
  • Description:
    Control slides, IHC, Treponema pallidum
  • Packaging:
    25/pack