Orders LinkContactUsLinkComponent
MERC Systems
MERC Systems
New Product
Supplier:   MOPEC
MERC Systems
Supplier:  MOPEC
New Product
Certificates
MERC Systems
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The patented Mopec MERC System® (Mortuary Enhanced Remains Cooling) is the first direct contact cooling system using liquid cooling technology to store human remains, and is the only human remains cooling system that is completely portable, scalable, and interoperable.
  • 0.75 to 2 ton cooling unit
  • 115 to 220 Volt/30 Amp/single phase
  • Stainless steel construction
  • Customer required to install the correct receptacle #L14-30R
The Mopec MERC System is one of the most efficient human remains cooling solutions on the market, and in many cases eliminates the need for large, stationary coolers, or refrigeration units. Available in three different unit sizes, storing 8, 24, and 48 bodies, based on storage cooling capacities with three case options for the cooling compressor unit.
Product Family Options
Product Information
  • Capacity
    Dimensions
    Availability
    Price
  • 78142-104
    MERC System
    MERC System
    8 body system
    27L×18W×32" H
    Log in for availability
    Log in for availability
    Each
    Each - $35,162.90
    $35,162.90
    Each
       $35,162.90
    Supplier Number: K-MERC08-SS
    Specifications:
    New Product
    New Product
  • 78142-105
    MERC System
    MERC System
    24 body system
    36L×22W×28" H
    Log in for availability
    Log in for availability
    Each
    Each - $63,907.37
    $63,907.37
    Each
       $63,907.37
    Supplier Number: K-MERC24-SS
    Specifications:
    New Product
    New Product
  • 78142-106
    MERC System
    MERC System
    48 body system
    43L×32W×39" H
    Log in for availability
    Log in for availability
    Each
    Each - $98,992.66
    $98,992.66
    Each
       $98,992.66
    Supplier Number: K-MERC48-SS
    Specifications:
    New Product