Order Entry
United States
Orders LinkContactUsLinkComponent
General Purpose Cleaning Solution

General Purpose Cleaning Solution

General purpose cleaning solution, 500 ml bottle
Catalog # 470357-992
Supplier:   HANNA INSTRUMENTS
Supplier #:  HI7061L
General Purpose Cleaning Solution
Catalog # 470357-992
Supplier:  HANNA INSTRUMENTS
CAS Number:  
General Purpose Cleaning Solution
General Purpose Cleaning Solution

General purpose cleaning solution, 500 ml bottle

CAS Number:  
Price
$22.99
Log in for availability
Each
Product Details & Documents

Gentle cleaning solution designed to remove buildup and maintain electrode accuracy.

  • Formulated to remove residues and contaminants from pH and ORP electrodes
  • Safe for use with glass, polymer, and combination sensors
  • Prevents inaccurate readings caused by sample buildup
  • Ready-to-use liquid requires no dilution
  • Airtight, tamper-proof bottle maintains solution stability
  • Long shelf life supports repeated use in classroom and lab environments

The General Purpose Cleaning Solution (500 mL) is specially formulated to clean pH and ORP electrodes without damaging sensitive glass membranes or reference junctions. Regular cleaning helps restore response time and measurement accuracy by removing contaminants such as salts, proteins, and organic deposits left behind from samples. This ready-to-use solution simplifies instrument maintenance and is safe for both glass and polymer-bodied sensors. Each bottle is sealed and labeled with a lot number and expiration date to ensure quality and traceability.

The solution is packaged in an airtight, tamper-evident HDPE container that preserves chemical stability during storage and transport. When stored properly, the solution maintains full effectiveness for up to five years unopened, making it ideal for educational and research laboratories that perform routine pH testing. Perfect for use in chemistry, biology, and environmental science courses, this cleaning solution supports consistent, accurate readings by ensuring electrodes remain free from buildup and ready for reliable calibration and measurement.

Specifications

Specifications

  • Description:
    General purpose cleaning solution, 500 ml bottle

Specifications