Order Entry
United States
Orders LinkContactUsLinkComponent
TMB EASE ready-to-use

TMB EASE ready-to-use

Glass bottle, 100 ml
Catalog # ALT494067100ML
New Product
Supplier:   Aladdin Scientific
Supplier #:  T494067-100ML
TMB EASE ready-to-use
Catalog # ALT494067100ML
Supplier:  Aladdin Scientific
New Product
CAS Number:  
TMB EASE ready-to-use

Glass bottle, 100 ml

CAS Number:  
Price
$329.00
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

TMB (3, 3', 5, 5'-tetramethylbenzidine) is a chromogenic substrate for Horseradish Peroxidase (HRP). TMB produces a deep blue color during the enzymatic degradation of hydrogen peroxide by HRP, and the addition of a stop solution gives a clear yellow color that absorbs at 450nm.TMB EASE characteristics are:1:1 replacement of carcinogenic 2mg OPD  Aqueous based – no organic solvents (REACH Annex XVII compliant)Low lot-to-lot variationProduct CharacteristicsTMB (3, 3', 5, 5'-tetramethylbenzidine) is a chromogenic substrate for Horseradish Peroxidase (HRP). TMB produces a deep blue color during the enzymatic degradation of hydrogen peroxide by HRP, and the addition of a stop solution gives a clear yellow color that absorbs at 450nm.TMB EASE is a ready-to-use and stable aqueous formulation. This substrate gives a slow assay reaction and is highly suitable for the direct switch from OPD (ortho-phenylenediamine dichloride) based assays. It does not contain NMP (1-methyl-2-pyrrolidone) making it REACH Restricted Substances List Annex XVII compliant, while ensuring maximal safety during use, and minimal waste problems after use.Composition and PropertiesReady-to-use substrate: Includes substrate buffer and hydrogen peroxide. No other reagents should be added.Working Procedure1. The desired amount of substrate is poured into a sealed container and allowed to reach room temperature in the dark before use.2. A recommended volume of 100 µl substrate solution per microtiter well is added. Time of development is typically 5-30 minutes. Keep the plate in the dark during incubation.3. In kinetic assays, the blue color is read directly at 650 nm. For endpoint assays 100 µl (equal volumes) of 0.20 M sulphuric acid is added and the absorbance is read at 450/620 nm. The yellow solution is stable for at least one hour.Tips and Tricks • If reduced intensity is required, it is recommended to shorten the time of development or to dilute the conjugate/antibody in the immunoassay further. • Dilution of the substrate is not recommended.• In case of incidents of precipitation with very high OD-values, we can recommend the use of the TMB X-tnd (cat. no. T494100). Handling and Storage • Store solution at 2-8 °C in the dark. • Avoid exposure to light, heat and contamination with metal ions or peroxidase. • Re-dispense only into bottles made of High Density Polyethylene (HDPE), amber color. • Dispensing guidelines are available upon request.

  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.