D-(-)-Mannitol, Supelco®
Glass bottle for solids, 50 g
78455-757
New Product
: 78513-50G
: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
- :69-65-8
- :HOH₂C(CH(OH))₄CH₂OH
- :MFCD00064287
- :182.17 g/mol
- :Ambient
- :200-711-8
- Pack type:Glass bottle for solids
- Size:50 g