Supelco® BioUltra Phosphate Buffer Solution
Phosphate buffer, >0.044 M Na₂HPO₄, >0.022 M NaH₂PO₄, 100 ml
Catalog # 78455-737
New Product
Supplier: MilliporeSigma
Supplier #: 76847-100ML-F
Supelco® BioUltra Phosphate Buffer Solution
Catalog # 78455-737
Supplier: MilliporeSigma
New Product
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Concentration:>0.044 M Na₂HPO₄, >0.022 M NaH₂PO₄
- Description:Phosphate buffer
- Size:100 ml
- Cat. No.:78455-737
- M-Clarity quality level:MQ100
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



