Orders LinkContactUsLinkComponent
Solid Door Wall Cabinets
Solid Door Wall Cabinets
New Product
Supplier:   MOPEC
Solid Door Wall Cabinets
Supplier:  MOPEC
New Product
Certificates
Solid Door Wall Cabinets
Solid Door Wall Cabinets
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Wall cabinet with two solid doors.
  • 304 stainless steel construction with a polished no. 4 finish
Wall cabinets vary in depth and height, this cabinet has solid doors.
Product Family Options
Product Information
  • Type
    Material
    Length
    Width
    Height
    Availability
    Price
  • 78142-194
    2 solid door wall cabinet
    2 solid door wall cabinet
    Storage
    Stainless Steel
    36"
    13"
    30.6"
    Log in for availability
    Log in for availability
    Each
    Each - $4,302.26
    $4,302.26
    Each
       $4,302.26
    Supplier Number: LC203-36
    Specifications:
    New Product
    New Product
  • 78142-195
    2 solid door wall cabinet
    2 solid door wall cabinet
    Storage
    Stainless Steel
    48"
    13"
    30.6"
    Log in for availability
    Log in for availability
    Each
    Each - $6,575.98
    $6,575.98
    Each
       $6,575.98
    Supplier Number: LC203-48
    Specifications:
    New Product