Orders LinkContactUsLinkComponent
Inside Corner Fillers
Inside Corner Fillers
New Product
Supplier:   MOPEC
Inside Corner Fillers
Supplier:  MOPEC
New Product
Certificates
Inside Corner Fillers
Inside Corner Fillers
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Inside corner filler for standing height casework.
  • Allows for seamless integration into lab workspaces, made for easy cleaning
Casework to store various items for labwork.
Product Family Options
Product Information
  • Material
    Width
    Length
    Height
    Availability
    Price
  • 78142-229
    Stationary, 1" long
    Stationary, 1" long
    Stainless steel
    1"
    1"
    35.1" (Standing height)
    Log in for availability
    Log in for availability
    Each
    Each - $414.24
    $414.24
    Each
       $414.24
    Supplier Number: LCFCH-01S
    Specifications:
    New Product
    New Product
  • 78142-230
    Stationary, adjustable
    Stationary, adjustable
    Stainless steel
    18"
    18"
    34.6" (Standing height)
    Log in for availability
    Log in for availability
    Each
    Each - $2,238.70
    $2,238.70
    Each
       $2,238.70
    Supplier Number: LCFCH-ADJS
    Specifications:
    New Product