Order Entry
United States
Orders LinkContactUsLinkComponent
Gentamicin sulfate USP

Gentamicin sulfate USP

New Product
Supplier:   G-Biosciences
Gentamicin sulfate USP
Supplier:  G-Biosciences
New Product
Certificates
Gentamicin sulfate USP

5 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Gentamicin efficiently inhibits growth of many Gram-positive and Gram-negative microorganisms. It does so by inhibiting protein synthesis by binding to the L6 protein of the 30S ribosomal subunit. It is also effective against mycoplasma. Recommended for use in cell culture applications.

  • USP Grade
Synonyms:  Gentamicin sulfate salt, NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
  • CAS Number:1405-41-0
  • Formula:C₂₁H₄₃N₅O₇.H₂SO₄
  • Molecular Weight:575.67 g/mol
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    Each
    Each - $708.80
    $708.80
    Each
       $708.80

    Supplier Number: RC-174
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Bottle
    10 g
    Log in for availability
    Log in for availability
    Each
    Each - $147.25
    $147.25
    Each
       $147.25

    Supplier Number: RC-173
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Vial
    1 g
    Log in for availability
    Log in for availability
    Each
    Each - $59.51
    $59.51
    Each
       $59.51

    Supplier Number: RC-172
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    Each
    Each - $217.78
    $217.78
    Each
       $217.78

    Supplier Number: RC-175
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    Each
    Each - $92.27
    $92.27
    Each
       $92.27

    Supplier Number: RC-176
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Bottle
    1 kg
    Discontinued
    Supplier Number: RC-859
    Specifications:
    New Product
    New Product
  • Bottle
    500 g
    Discontinued
    Supplier Number: RC-858
    Specifications:
    New Product
    New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.