Order Entry
United States
Orders LinkContactUsLinkComponent
Chlorophenol red

Chlorophenol red

Bottle, 50 g
Catalog # 76286-210
Supplier:   G-Biosciences
Supplier #:  RC-649
Chlorophenol red
Catalog # 76286-210
Supplier:  G-Biosciences
CAS Number:  4430-20-0
Chlorophenol red

Bottle, 50 g

CAS Number:  4430-20-0
Price
$597.65
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Chlorophenol red is an indicator dye that changes color from yellow to violet in the pH range 4.8 to 6.7.

  • Molecular Biology Grade Brown crystalline powder
Synonyms:  3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide
  • CAS Number:4430-20-0
  • Formula:C₁₉H₁₂Cl₂O₅S
  • Molecular Weight:423.27 g/mol
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    50 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.