Fecal Collection Tubes
Fecal collection tubes, Orange, Screw cap, 10 Tubes/pack
Catalog # 77902-068
New Product
Supplier: G-Biosciences
Supplier #: 786-2184
Jump to:
Product Details & Documents
Fecal collection tube is designed for the collection and preservation of DNA/RNA from stool specimens. Fecal collection tube is supplied with screwcap attached with spoon.
Specifications
- Description:Fecal collection tubes
- Cap color:Orange
- Closure type:Screw cap
- Packaging:10 Tubes/pack