Order Entry
United States
Orders LinkContactUsLinkComponent
Lead subacetate 33,0% (by titrimetric analysis) ACS

Lead subacetate 33,0% (by titrimetric analysis) ACS

New Product
Lead subacetate 33,0% (by titrimetric analysis) ACS
New Product
Certificates
Lead subacetate 33,0% (by titrimetric analysis) ACS
Lead subacetate 33,0% (by titrimetric analysis) ACS

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Store at RT.

Synonyms:  Horne′s compound, Lead(II) hydroxide acetate, Acetic acid lead (II) salt basic, Lead subacetate
  • CAS Number:1335-32-6
  • Formula:Pb(C₂H₃O₂)₂·2Pb(OH)₂
  • MDL Number:MFCD00013041
  • Molecular Weight:807.72 g/mol
  • Storage Temperature:Room Temperature
  • UN:1616
Specifications for Product Family

Specification Test Results

  • Assay by titration:
    ≥33.0% (PbO)
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $23.76
    $23.76
    Each(1/Itm/Bx)
       $23.76

    Supplier Number: 22888-100G
    Specifications:
    New Product
    New Product
  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $108.22
    $108.22
    Each(1/Itm/Bx)
       $108.22

    Supplier Number: 22888-1KG
    Specifications:
    New Product
    New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.