Order Entry
United States
Orders LinkContactUsLinkComponent
Cycloheximide (from Streptomyces griseus) ≥98% (by HPLC)

Cycloheximide (from Streptomyces griseus) ≥98% (by HPLC)

New Product
Cycloheximide (from Streptomyces griseus) ≥98% (by HPLC)
New Product
Certificates
Cycloheximide (from Streptomyces griseus) ≥98% (by HPLC)
Cycloheximide (from Streptomyces griseus) ≥98% (by HPLC)

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Cycloheximide, derived from the bacterium Streptomyces griseus, is a potent inhibitor of protein synthesis in eukaryotic cells, making it an invaluable tool in molecular biology and biochemistry research. This compound is widely utilized in studies involving gene expression, cellular signaling, and the effects of various compounds on protein synthesis. Its ability to selectively inhibit the elongation phase of translation allows researchers to dissect the roles of specific proteins in cellular processes, providing insights into mechanisms of action for drugs and potential therapeutic targets.

In addition to its research applications, Cycloheximide has been employed in agricultural settings to study plant responses to stress and pathogen resistance, showcasing its versatility across different scientific fields. The compound's unique properties, including its effectiveness at low concentrations and its ability to penetrate cell membranes, enhance its utility in both laboratory and field studies. Researchers and industry professionals can leverage Cycloheximide to advance their understanding of complex biological systems and develop innovative solutions in health and agriculture.

Store at 0 to 8 °C.

Synonyms:  Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
  • CAS Number:66-81-9
  • Formula:C₁₅H₂₃NO₄
  • MDL Number:MFCD00082346
  • Molecular Weight:281.35 g/mol
  • Storage Temperature:Refrigerator
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic bottle
    1 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $45.50
    $45.50
    Each(1/Itm/Bx)
       $45.50

    Supplier Number: 00083-1G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product
  • Plastic bottle
    5 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $171.87
    $171.87
    Each(1/Itm/Bx)
       $171.87

    Supplier Number: 00083-5G
    Specifications:
    New Product
    New Product
  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $2,203.98
    $2,203.98
    Each(1/Itm/Bx)
       $2,203.98

    Supplier Number: 00083-100G
    Specifications:
    New Product
    New Product
  • Plastic bottle
    25 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $697.59
    $697.59
    Each(1/Itm/Bx)
       $697.59

    Supplier Number: 00083-25G
    Specifications:
    New Product
    New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.